Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ricinoleic Acid, MP Biomedicals™

Catalog No. 0219151490 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-191-51490 500 mg
ICN19151480 100 mg
ICN19151491 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-191-51490 Supplier MP Biomedicals, Inc Supplier No. 0219151490
Add to Cart
Add to Cart

Ricinoleic Acid is used as a substrate for the synthesis of the conjugated linoleic acids.

  • Ricinoleic Acid is a naturally occurring 12-hydroxy fatty acid constituting about 90% of the fatty acids in castor oil.
  • Has the capacity to serve as a substrate for the synthesis of the conjugated linoleic acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-22-0
Molecular Formula C18H34O3
Molecular Weight (g/mol) 298.47
InChI Key WBHHMMIMDMUBKC-QJWNTBNXSA-N
IUPAC Name (9Z,12R)-12-hydroxyoctadec-9-enoic acid
SMILES CCCCCC[C@@H](O)C\C=C/CCCCCCCC(O)=O

Specifications

Melting Point 5.5°C
Color Colorless
Density 0.940 g/mL at 27.4°C/4°C
Boiling Point 245°C
Assay Percent Range >99%
Quantity 500 mg
Solubility Information Water Solubility: 3460 mg/L at 25°C)
Formula Weight 298.5
Vapor Pressure 4.49X10-3 mm Hg at 25°C
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Ricinoleic Acid
Show More Show Less
EINECSNumber 205-470-2

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.