Learn More
Cayman Chemical Ricinoleic Acid ethyl ester, Purity: ≥98%, CAS: 55066-53-0, 500 mg
SDP
Catalog No. 502891196
Encompass_Preferred
A stabilized, lipid-soluble form of the free acid that has been used to produce biofuels as well as skin-conditioning agents, emulsion stabilizers, and surfactants in cosmetics
Catalog No. 50-289-1196 Supplier Cayman Chemical Supplier No. 10009730500 MG
A stabilized, lipid-soluble form of the free acid that has been used to produce biofuels as well as skin-conditioning agents, emulsion stabilizers, and surfactants in cosmetics