Learn More
Cayman Chemical Ricinoleic Acid ethyl ester
SDP
Catalog No. 502891195
Encompass_Preferred
A stabilized, lipid-soluble form of the free acid that has been used to produce biofuels as well as skin-conditioning agents, emulsion stabilizers, and surfactants in cosmetics
Catalog No. 50-289-1195 Supplier Cayman Chemical Supplier No. 100097301 G
Add to Cart
Add to Cart
A stabilized, lipid-soluble form of the free acid that has been used to produce biofuels as well as skin-conditioning agents, emulsion stabilizers, and surfactants in cosmetics