Learn More
Cayman Chemical Ricinoleic Acid Fatty Acids an
SDP
Catalog No. 502889357
Encompass_Preferred
Ricinoleic acid is a naturally occurring 12-hydroxy fatty acid. It constitutes about 90% of the fatty acids in castor oil. Ricinoleic acid can serve as a substrate for the synthesis of conjugated linoleic acids. It is considered safe and non-toxic as a food constituent in humans, up to a daily intake of 2.5 grams per day. In high doses, castor oil has a laxative effect.
Catalog No. 50-288-9357 Supplier Cayman Chemical Supplier No. 100093695 G
Add to Cart
Add to Cart
Ricinoleic acid is a naturally occurring 12-hydroxy fatty acid. It constitutes about 90% of the fatty acids in castor oil. Ricinoleic acid can serve as a substrate for the synthesis of conjugated linoleic acids. It is considered safe and non-toxic as a food constituent in humans, up to a daily intake of 2.5 grams per day. In high doses, castor oil has a laxative effect.