Learn More
Cayman Chemical Ricinoleic Acid methyl ester, Purity: ≥98%, CAS: 141-24-2, 1 g
SDP
Catalog No. 502889732
Encompass_Preferred
Ricinoleic acid methyl ester is a neutral, more lipophilic form of the free acid that can be used as an analytical standard for ricinoleic acid.
Catalog No. 50-288-9732 Supplier Cayman Chemical Supplier No. 100097291 G
Ricinoleic acid methyl ester is a neutral, more lipophilic form of the free acid that can be used as an analytical standard for ricinoleic acid.