Learn More
Enzo Life Sciences RICK polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADIAAP460E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of human RICK. Species reactivity: Human, Mouse, Rat. Applications: ICC, IP, WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-7165