Learn More
Bioss Rickettsia
SDP
Catalog No. 5000065686
Encompass_Preferred
Rickettsia Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA IHC Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide
Catalog No. 50-000-65686 Supplier Bioss Supplier No. BSM72387M
Rickettsia Primary Antibody Unconjugated Host Species Mouse Target Species Clonality Monoclonal Applications ELISA IHC Concentration 100 g/ml Size 100 ug Storage Buffer PBS (pH 7 2) with 0 1% azide