Learn More
Enzo Life Sciences Rictor monoclonal antibody (1G11) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905766100
Encompass_Preferred
Clone: 1G11. Host: Mouse. Isotype: IgG1. Immunogen: Synthetic peptide corresponding to a portion of Rictor. UniProt ID: Q6R327. Species reactivity: Human, Mouse, Rat. Applications: WB. Recommended Dilutions: Western Blot (0.5µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Thiophilic adsorption and size exclusion chromatography purified. Formulation: Lyophilized from 1ml of 2x PBS containing 0.09% sodium azide, PEG, and sucrose. Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and store at -80°C.
Catalog No. 50-200-7043