Learn More
Selleck Chemical LLC Ridaforolimus (Deforolimus, MK-8669) 10mM/1mL 572924-54-0 AP23573
SDP
Catalog No. 501366112
Encompass_Preferred
Ridaforolimus (Deforolimus, MK-8669, AP23573) is a selective mTOR inhibitor with IC50 of 0.2 nM in HT-1080 cell line; while not classified as a prodrug, mTOR inhibition and FKBP12 binding is similar to rapamycin. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6112 Supplier Selleck Chemical LLC Supplier No. S102210MM/1ML
Ridaforolimus (Deforolimus, MK-8669, AP23573) is a selective mTOR inhibitor with IC50 of 0.2 nM in HT-1080 cell line; while not classified as a prodrug, mTOR inhibition and FKBP12 binding is similar to rapamycin. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.