Learn More
R Biopharm Inc QUICK Gliadin, 25 test strips
Catalog No. NC0730224
Encompass
RIDA QUICK Gliadin (single packaged) can be used for gluten detection on surfaces for hygiene control (swab test), in cleansing waters, and for the qualitative detection of gliadin / gluten in raw material and processed food. The test has been developed for the detection of low amounts of gluten (contamination). No high-dose-hook-effect is observed at high concentrations.
Catalog No. NC0730224 Supplier R Biopharm Inc Supplier No. R7004
RIDA QUICK Gliadin (single packaged) can be used for gluten detection on surfaces for hygiene control (swab test), in cleansing waters, and for the qualitative detection of gliadin / gluten in raw material and processed food. The test has been developed for the detection of low amounts of gluten (contamination). No high-dose-hook-effect is observed at high concentrations.