Learn More
R Biopharm Inc RIDASCREEN GLIADIN
Catalog No. NC0573722
Encompass
A sandwich enzyme immunoassay for the quantitative analysis of contaminations by prolamins from wheat, rye, & barley in raw products like flours & spices and in processed food. Microtiter plate (12 strips w/8 removable wells ea)
Catalog No. NC0573722 Supplier R Biopharm Inc Supplier No. R7001
A sandwich enzyme immunoassay for the quantitative analysis of contaminations by prolamins from wheat, rye, & barley in raw products like flours & spices and in processed food. Microtiter plate (12 strips w/8 removable wells ea)