Learn More
MSC Karcher 71 inch Single Brush Sweeper, Self Propelled, 81 gal.
Catalog No. 502818140
Encompass
This industrial sweeper is designed for efficient floor cleaning. It features a self-propelled mechanism and a wide sweeping path, making it suitable for large areas.
  • Self propelled
  • Rubber wheels
  • Polyethylene bristles
  • Capacity of 81 gal.
  • Sweeping width of 71 inch
  • Single brush sweeping mechanism
Catalog No. 50-281-8140 Supplier MSC Supplier No. 16619322
Add to Cart
Add to Cart
Request a Quote
This industrial sweeper is designed for efficient floor cleaning. It features a self-propelled mechanism and a wide sweeping path, making it suitable for large areas.
  • Self propelled
  • Rubber wheels
  • Polyethylene bristles
  • Capacity of 81 gal.
  • Sweeping width of 71 inch
  • Single brush sweeping mechanism