Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Karcher KM 150/500 R LPG, LPG, 1 ea

Catalog No. 502819391
Encompass

The Kärcher KM 150/500 R LPG is an LPG-powered ride-on vacuum sweeper for commercial and heavy-duty industrial applications. Featuring a robust steel chassis, low noise, and exhaust-free emissions, it suits both indoor and outdoor use. It efficiently vacuums fine and coarse dirt, with an automatic closing waste container for enhanced cleanliness.

  • Robust steel chassis
  • Three-wheel rear steering
  • Low noise, exhaust-free emissions
  • Dustpan principle for fine and coarse dirt
  • Automatic closing waste container
  • Roller brush auto-adjusts, tool-free replacement
  • Dual flat fold filters for clean air, push-button cleaning
  • EASY Operation Concept control
  • Working width with 1 side brush: 59 in
  • Area performance: 193,752 ft²/hr
  • Waste container capacity: 132 gal
  • Dimensions (L x W x H): 96 x 61 x 65 in
  • Power source: Propane

Catalog No. 50-281-9391 Supplier MSC Supplier No. 16619009
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The Kärcher KM 150/500 R LPG is an LPG-powered ride-on vacuum sweeper for commercial and heavy-duty industrial applications. Featuring a robust steel chassis, low noise, and exhaust-free emissions, it suits both indoor and outdoor use. It efficiently vacuums fine and coarse dirt, with an automatic closing waste container for enhanced cleanliness.

  • Robust steel chassis
  • Three-wheel rear steering
  • Low noise, exhaust-free emissions
  • Dustpan principle for fine and coarse dirt
  • Automatic closing waste container
  • Roller brush auto-adjusts, tool-free replacement
  • Dual flat fold filters for clean air, push-button cleaning
  • EASY Operation Concept control
  • Working width with 1 side brush: 59 in
  • Area performance: 193,752 ft²/hr
  • Waste container capacity: 132 gal
  • Dimensions (L x W x H): 96 x 61 x 65 in
  • Power source: Propane

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.