Learn More
MSC Karcher Ride-On Sweeper, LPG-Powered, Overhead Guard, 1 ea
Catalog No. 502817875
Encompass
The KM 130/300 R LPG with overhead guard is a robust, fully hydraulic, LP-powered industrial sweeper designed for extreme cleaning tasks.
  • Utilizes dustpan principle to vacuum fine sweepings and coarse dirt
  • Roller brush automatically adjusts to uneven surfaces
  • Innovative sweeping system minimizes wear and tear
  • Tool-free main roller brush change
  • Waste container automatically closes during transport
  • Two horizontally installed flat pleated filters ensure clean air
  • Filter cleaning via push-button with dual scraper
  • Optimally accessible filter for easy, tool-free changes
  • Easy access to the motor
  • EASY Operation Concept for convenient function selection
Catalog No. 50-281-7875 Supplier MSC Supplier No. 16619363
Add to Cart
Add to Cart
Request a Quote
The KM 130/300 R LPG with overhead guard is a robust, fully hydraulic, LP-powered industrial sweeper designed for extreme cleaning tasks.
  • Utilizes dustpan principle to vacuum fine sweepings and coarse dirt
  • Roller brush automatically adjusts to uneven surfaces
  • Innovative sweeping system minimizes wear and tear
  • Tool-free main roller brush change
  • Waste container automatically closes during transport
  • Two horizontally installed flat pleated filters ensure clean air
  • Filter cleaning via push-button with dual scraper
  • Optimally accessible filter for easy, tool-free changes
  • Easy access to the motor
  • EASY Operation Concept for convenient function selection