Learn More
Chemglass Life Sciences Universal Ridge Kwik Klamp II
SDP
Catalog No. 50871982
Encompass_Preferred
Universal Ridge Kwik Klamp II. Universal Kwik Klamp II is constructed of aluminum that has a chemically resistant Hardcote and PTFE anodized finish. Versatile design can be used on 1/2” OD Rod. Useful in attaching temperature monitors or controllers, additional supports and clamps. The optional CG-3498-03 1/2” OD rod, bent 90° is necessary when supporting a temperature monitor or controller to a reactor stand or laboratory hood lattice.Features:• Heavy Duty Design• Corrosion Resistant• Large Clamping Knobs• Universal Design Securely Fastens To 10L Through 100L Reactor Frames And Stationary Or Mobile Benchtop Stands• 1/2" Diameter Rod Commonly Used For Lattice In Hoods• Clamps and Support Rods Can Be Mounted Horizontally Or Vertically• Secure Up To Four Different Rods, Clamps Or SupportsRidge Klamp for use with: CG-1968-X Support Frame
Catalog No. 50-871-982 Supplier Chemglass Life Sciences Supplier No. CG925330
Universal Ridge Kwik Klamp II. Universal Kwik Klamp II is constructed of aluminum that has a chemically resistant Hardcote and PTFE anodized finish. Versatile design can be used on 1/2” OD Rod. Useful in attaching temperature monitors or controllers, additional supports and clamps. The optional CG-3498-03 1/2” OD rod, bent 90° is necessary when supporting a temperature monitor or controller to a reactor stand or laboratory hood lattice.Features:• Heavy Duty Design• Corrosion Resistant• Large Clamping Knobs• Universal Design Securely Fastens To 10L Through 100L Reactor Frames And Stationary Or Mobile Benchtop Stands• 1/2" Diameter Rod Commonly Used For Lattice In Hoods• Clamps and Support Rods Can Be Mounted Horizontally Or Vertically• Secure Up To Four Different Rods, Clamps Or SupportsRidge Klamp for use with: CG-1968-X Support Frame