Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Ridgeline™ 4-Point Vented Capstyle Hard Hat

Catalog No. 19155434 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Blue
Gray
Hi-vis Lime
Hi-vis Orange
Matte Black Graphite
Red
White
White Graphite
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-155-434 White
19-155-436 Gray
19-155-439 White Graphite
19-155-441 Matte Black Graphite
19-155-444 Red
19-155-446 Yellow
19-155-448 Hi-vis Lime
19-155-451 Hi-vis Orange
19-155-453 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-155-434 Supplier Pyramex™ Supplier No. HP44110V
Add to Cart
Add to Cart

Ridgeline™ 4-Point Vented Capstyle Hard Hat is strong, yet ultra-light for seemingly weightless protection.

  • ABS material – strong, yet ultra-light for seemingly weightless protection
  • Low profile design – offers a low center of gravity for better balance
  • Rear padded suspension – provides additional level of comfort on the wearers' neck
  • 4-position harness points allows you to move the harness forward, backward, up or down – allowing for the most optimal position for ultimate comfort
  • Replaceable sweat band – padded fabric and polyurethane foam for all day wear
  • Vented pressure pad keeps the straps in place providing an additional level of comfort
  • Easy to grasp knob adjusts the suspension from 6 1/2 to 8

Specifications

Certifications/Compliance ANSI Z89.1 standards, Type 1 --- Class C, G
Color White
For Use With (Application) Construction
Material Shell: ABS, Suspension: HDPE and high impact Nylon, Sweat band: Polyester Fabric and Polyurethane Foam
Product Type Vented 4-Point Ratchet
Size 6.5 to 8
Weight (Metric) 358 g
Ventilated Vented
Head Style Short Brim (Cap Style)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.