Learn More
Pyramex Safety Products Ridgeline Cap Style 4-Point Ratchet for CSA
Shop All Pyramex Safety Products ProductsDescription
- Made from ABS material, this thermoplastic resin is a lightweight material that combines strength and rigidity.
- Rear padded suspension provides additional level of comfort on the wearers' neck.
- Vented pressure pad keeps the straps in place providing an additional level of comfort.
- Easy to grasp triangle knob adjusts the suspension from 6.5 to 8-inch hat sizes.
- Replaceable sweat band padded fabric and polyurethane foam for all day wear.
- 4-position harness points allows users to move the harness forward, backward, up, or down-allowing for the most optimal position for ultimate comfort.
Specifications
Specifications
| Color | Blue |
| Material | ABS |
| Product Type | 4-point ratchet |
| Size | Adjustable (6.5 to 8) |
| Ventilated | Ventilated |