Learn More
Pyramex Safety Products Ridgeline Cap Style Hard Hat - 4 Point Ratchet Suspension
Catalog No. 19555097 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Size:
Adjustable (6.5 to 8)
Catalog No. Size
19-555-097 Adjustable (6.5 to 8)
1 options
Catalog No. 19-555-097 Supplier Pyramex Safety Products Supplier No. HP44170
Add to Cart
Add to Cart
Pyramex Safety™ Ridgeline Cap Style Hard Hat - 4 Point Ratchet Suspension features a low profile design and low center of gravity for better balance.
  • Made from ABS material, this thermoplastic resin is a lightweight material that combines strength and rigidity.
  • Rear padded suspension provides additional level of comfort on the wearers' neck.
  • Vented pressure pad keeps the straps in place providing an additional level of comfort.
  • Easy to grasp triangle knob adjusts the suspension from 6.5 to 8-in. hat sizes.
  • Replaceable sweat band padded fabric and polyurethane foam for all day wear.
  • 4-position harness points allows users to move the harness forward, backward, up or down-allowing for the most optimal position for ultimate comfort.
Specifications
Color Hi-vis Pink
Product Type 4-point Ratchet Suspension
Size Adjustable (6.5 to 8)