Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Ridgeline™ Capstyle Hard Hat

Catalog No. 19155433 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Product Type:
4-Point Ratchet
6-Point Ratchet
Color:
Blue
Gray
Green
Hi-vis Lime
Hi-vis Orange
Light Blue
Red
Slate Gray
White
Yellow
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Color
19-155-433 4-Point Ratchet White
19-155-435 4-Point Ratchet Gray
19-155-437 4-Point Ratchet Slate Gray
19-155-443 4-Point Ratchet Red
19-155-445 4-Point Ratchet Yellow
19-155-447 4-Point Ratchet Hi-vis Lime
19-155-449 4-Point Ratchet Green
19-155-450 4-Point Ratchet Hi-vis Orange
19-155-452 4-Point Ratchet Blue
19-155-454 4-Point Ratchet Light Blue
19-155-455 6-Point Ratchet White
19-155-456 6-Point Ratchet Red
19-155-457 6-Point Ratchet Yellow
19-155-458 6-Point Ratchet Hi-vis Lime
19-155-459 6-Point Ratchet Green
19-155-460 6-Point Ratchet Hi-vis Orange
19-155-461 6-Point Ratchet Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
Catalog No. 19-155-433 Supplier Pyramex™ Supplier No. HP44110
Add to Cart
Add to Cart

Ridgeline™ Capstyle Hard Hat is strong, yet ultra-light for seemingly weightless protection.

  • ABS material – strong, yet ultra-light for seemingly weightless protection
  • Low profile design – offers a low center of gravity for better balance
  • Rear padded suspension – provides additional level of comfort on the wearers’ neck
  • 4-position harness points allows you to move the harness forward, backward, up or down – allowing for the most optimal position for
    ultimate comfort
  • Replaceable sweat band – padded fabric and polyurethane foam for all day wear
  • Vented pressure pad keeps the straps in place providing an additional level of comfort
  • Easy to grasp knob adjusts the suspension from 6 1/2 to 8

Specifications

Certifications/Compliance ANSI Z89.1 standards, Type 1 --- Class C, G, and E
Color White
For Use With (Application) Construction
Liner Material Polyester Fabric and Polyurethane Foam
Material Shell: ABS, Suspension: HDPE and high impact Nylon, Sweat band: Polyester Fabric and Polyurethane Foam
Product Type 4-Point Ratchet
Size 6.5 to 8
Weight (Metric) 358 g
Ventilated Not Vented
Head Style Short Brim (Cap Style)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.