Learn More
Pyramex Safety Products Ridgeline Full Brim 6-Point Ratchet Suspension
Catalog No. 19555112 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-555-112 Blue
19-555-108 White
19-555-109 Red
19-555-110 Yellow
19-555-111 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-555-112 Supplier Pyramex Safety Products Supplier No. HP56160
Add to Cart
Add to Cart
Pyramex Safety™ Ridgeline Full Brim 6-Point Ratchet Suspension features a low profile design and low center of gravity for better balance.
  • Made from ABS material, this thermoplastic resin is a lightweight material that combines strength and rigidity.
  • Rear padded suspension provides additional level of comfort on the wearers' neck.
  • Vented pressure pad keeps the straps in place providing an additional level of comfort.
  • Easy to grasp triangle knob adjusts the suspension from 6.5 to 8-in. hat sizes.
  • Replaceable sweat band padded fabric and polyurethane foam for all day wear.
  • 4-position harness points allows users to move the harness forward, backward, up, or down-allowing for the most optimal position for ultimate comfort.
Specifications
Color Blue
Material ABS
Size Adjustable (6.5 to 8)
Ventilated Ventilated