Learn More
Pyramex™ Ridgeline™ Full Brim 6 Point Suspension Strap
Catalog No. 19155478 Shop All Pyramex Safety Products Products
Change view
Click to view available options
For Use With (Equipment):
Ridgeline Full Brim Hard Hats
Catalog No. For Use With (Equipment)
19-155-478 Ridgeline Full Brim Hard Hats
1 options
Catalog No. 19-155-478 Supplier Pyramex™ Supplier No. HP6PTSUS
Add to Cart
Add to Cart
Used to convert a 4-Point suspension to a 6-Point suspension for all Ridgeline full brim hard hats
Convert 4-point suspension to 6-point suspension
Specifications
For Use With (Equipment) Ridgeline Full Brim Hard Hats