Learn More
MSC Pyramex Full Brim Hard Hat, Ridgeline, 6-Point Ratchet, 1 ea.
Catalog No. 5000688296
Encompass
The full brim hat is designed to provide reliable head protection in various industrial environments. Engineered for durability and comfort, this protective headgear offers extended coverage for the wearer.
  • Features a full brim design for added protection from the elements
  • Includes a 6-point ratchet suspension system for a secure and comfortable fit
  • Equipped with Mips® safety system
Catalog No. 50-006-88296 Supplier MSC Supplier No. 88466313
The full brim hat is designed to provide reliable head protection in various industrial environments. Engineered for durability and comfort, this protective headgear offers extended coverage for the wearer.
  • Features a full brim design for added protection from the elements
  • Includes a 6-point ratchet suspension system for a secure and comfortable fit
  • Equipped with Mips® safety system