Learn More
Pyramex™ Ridgeline™ HGBR Ratchet Headgear
Catalog No. 19155360 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Material:
ABS, Nylon
Catalog No. Material
19-155-360 ABS, Nylon
1 options
Catalog No. 19-155-360 Supplier Pyramex™ Supplier No. HGBR
Add to Cart
Add to Cart
Ridgeline™ ratchet headgear
  • Constructed from durable nylon
  • Ratchet adjustment for easy fit
  • Pivoting action allows user to lift face shield while wearing
  • Adjustable pegs will accommodate a wide variety of face shields
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Color Black
    Includes Ratchet and pivoting action
    Material ABS, Nylon
    Size 6.25 in. (Crown Depth)
    Suspension Type Adjustable ratchet suspension, Adjustable pin lock for top of head, Padded brow guard
    Weight (English) 7.4 oz.
    Weight (Metric) 210 g