Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex™ Ridgeline™ HHABCMR Headgear Adapter

Catalog No. 19155365 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Material:
Nylon, ABS
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-155-365 Nylon, ABS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-155-365 Supplier Pyramex™ Supplier No. HHABCMR
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cap Style hard hat adapter

  • Dielectric
  • Hard hat adapters fit into slots of Ridgeline cap style hard hats
  • Can be used with cap mounted earmuff
  • Adjustable while using cap mounted earmuff
  • Fits Ridgeline cap style hard hats and accommodates a variety of face shields
  • Specifications

    Certifications/Compliance ANSI Z87.1
    Color Black
    For Use With (Equipment) Rounded, Cap-style Hard Hats
    Material Nylon, ABS
    Thickness 2.1 to 2.9 mm
    Weight (English) 3.17 oz.
    Weight (Metric) 90 g
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.