Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pyramex Safety Products Ridgeline™ Vented Full Brim 4 Point Ratchet Suspension Hard Hat

Catalog No. 19555105 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Size:
6.5 to 8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-555-105 6.5 to 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-555-105 Supplier Pyramex Safety Products Supplier No. HP54117V
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Matte black graphite pattern Ridgeline™ Vented Full Brim 4 Point Ratchet Suspension Hard Hat.

  • Pyramex Safety - Ridgeline Hard Hat - Graphite-Ridgeline Full Brim 4 Pt Ratchet Suspension
  • The Ridgeline full brim has a shell constructed from ABS materials.
  • Ratchet suspension is easy to adjust and allows the wearer to modify the fit while wearing hard hat.
  • Soft brow pad is replaceable.
  • Replaceable suspension and headband also available.
  • Vented.

Specifications

Color Black
For Use With (Application) Construction
Product Type 4-point Ratchet Suspension
Size 6.5 to 8
Ventilated Ventilated
Head Style Full Brim
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.