Learn More
Medchemexpress LLC HY-126972A 5mg Medchemexpress, RI(dl)-2 (TFA) CAS: Purity:>98%
SDP
Catalog No. 502081122
Encompass_Preferred
Medchemexpress, HY-126972A 5mg RI(dl)-2 (TFA) CAS: RI(dl)-2 TFA is a potent and selective RAD51-mediated D-loop formation inhibitor with an IC50 of 11.1 μM. RI(dl)-2 TFA does not influence RAD51 binding to ssDNA and inhibits homologous recombination (HR) activity in human cells (IC50 of 3.0 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1122 Supplier Medchemexpress LLC Supplier No. HY126972A5MG
Medchemexpress, HY-126972A 5mg RI(dl)-2 (TFA) CAS: RI(dl)-2 TFA is a potent and selective RAD51-mediated D-loop formation inhibitor with an IC50 of 11.1 μM. RI(dl)-2 TFA does not influence RAD51 binding to ssDNA and inhibits homologous recombination (HR) activity in human cells (IC50 of 3.0 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.