Learn More
Medchemexpress LLC RIDR-PI-103 | 2581114-71-6 | 98.0% | 511.53 g/mol | C27H25N7O4 | 5 MG
SDP
Catalog No. 5000221297
Encompass_Preferred
RIDR-PI-103 is a reactive oxygen species (ROS)-activated prodrug designed to release a pan-PI3K inhibitor upon ROS activation. Supplied as a solid research compound, it is intended for use in cellular and biochemical studies investigating ROS-dependent modulation of the PI3K pathway.
  • ROS-activated prodrug mechanism enabling conditional release of active inhibitor.
  • Releases a pan-PI3K inhibitory moiety upon oxidative activation.
  • High reported purity suitable for research applications.
  • Solid form for straightforward storage and formulation.
  • Available in small research pack sizes and DMSO solutions for immediate use.
Catalog No. 50-002-21297 Supplier Medchemexpress LLC Supplier No. HY1448765MG
RIDR-PI-103 is a reactive oxygen species (ROS)-activated prodrug designed to release a pan-PI3K inhibitor upon ROS activation. Supplied as a solid research compound, it is intended for use in cellular and biochemical studies investigating ROS-dependent modulation of the PI3K pathway.
  • ROS-activated prodrug mechanism enabling conditional release of active inhibitor.
  • Releases a pan-PI3K inhibitory moiety upon oxidative activation.
  • High reported purity suitable for research applications.
  • Solid form for straightforward storage and formulation.
  • Available in small research pack sizes and DMSO solutions for immediate use.