Learn More
Medchemexpress LLC RIDR-PI-103 | 2581114-71-6 | 98.0% | C27H25N7O4 | 10MG
SDP
Catalog No. 5000258503
Encompass_Preferred
RIDR-PI-103 is a reactive oxygen species (ROS)-activated prodrug that releases a pan-PI3K inhibitor upon activation. It is supplied for research use to study PI3K/mTOR signaling, ROS-dependent prodrug activation, and related cellular models. Available as a solid powder and as DMSO solutions for in vitro applications.
  • Reactive oxygen species-activated prodrug enabling controlled release under oxidative conditions.
  • Designed to study PI3K/mTOR signaling and ROS-dependent therapeutic strategies.
  • Available as solid powder and as ready-to-use DMSO solution for assay flexibility.
  • High purity suitable for biochemical and cellular assays.
  • Molecular formula C27H25N7O4; molecular weight 511.53 g/mol.
Catalog No. 50-002-58503 Supplier Medchemexpress LLC Supplier No. HY14487610MG
RIDR-PI-103 is a reactive oxygen species (ROS)-activated prodrug that releases a pan-PI3K inhibitor upon activation. It is supplied for research use to study PI3K/mTOR signaling, ROS-dependent prodrug activation, and related cellular models. Available as a solid powder and as DMSO solutions for in vitro applications.
  • Reactive oxygen species-activated prodrug enabling controlled release under oxidative conditions.
  • Designed to study PI3K/mTOR signaling and ROS-dependent therapeutic strategies.
  • Available as solid powder and as ready-to-use DMSO solution for assay flexibility.
  • High purity suitable for biochemical and cellular assays.
  • Molecular formula C27H25N7O4; molecular weight 511.53 g/mol.