Learn More
Medchemexpress LLC RIDR-PI-103 | 2581114-71-6 | 98.0% | C27H25N7O4 | 25 MG
SDP
Catalog No. 5000356586
Encompass_Preferred
RIDR-PI-103 is a reactive oxygen species (ROS)-induced agent release proagent. It features a self-cyclizing moiety linked to a pan-PI3K inhibitor (PI-103). It has been observed that Doxorubicin and RIDR-PI-103 exhibit a synergistic effect in MDA-MB-361 and MDA-MB-231 cells, leading to the inhibition of cancer cell proliferation. This product is intended for research use only.
  • Reactive oxygen species (ROS)-induced agent release proagent
  • Features a self-cyclizing moiety linked to a pan-PI3K inhibitor (PI-103)
  • Exhibits synergistic effect with Doxorubicin in MDA-MB-361 and MDA-MB-231 cells to inhibit cancer cell proliferation
  • Targets reactive oxygen species
  • Targets immunology/inflammation pathway
  • Targets metabolic enzyme/protease pathway
  • Targets NF-κB pathway
Catalog No. 50-003-56586 Supplier Medchemexpress LLC Supplier No. 25MGHY144876
RIDR-PI-103 is a reactive oxygen species (ROS)-induced agent release proagent. It features a self-cyclizing moiety linked to a pan-PI3K inhibitor (PI-103). It has been observed that Doxorubicin and RIDR-PI-103 exhibit a synergistic effect in MDA-MB-361 and MDA-MB-231 cells, leading to the inhibition of cancer cell proliferation. This product is intended for research use only.
  • Reactive oxygen species (ROS)-induced agent release proagent
  • Features a self-cyclizing moiety linked to a pan-PI3K inhibitor (PI-103)
  • Exhibits synergistic effect with Doxorubicin in MDA-MB-361 and MDA-MB-231 cells to inhibit cancer cell proliferation
  • Targets reactive oxygen species
  • Targets immunology/inflammation pathway
  • Targets metabolic enzyme/protease pathway
  • Targets NF-κB pathway