Learn More
Medchemexpress LLC Rifabutin | 72559-06-9 | 99.15% | 847.00 | 1 ML
SDP
Catalog No. 5000384494
Encompass_Preferred
Rifabutin (Ansamycin) is a semisynthetic ansamycin antibiotic with potent antimycobacterial properties, primarily used as a bactericidal antibiotic to treat tuberculosis. It inhibits DNA-dependent RNA polymerase and is effective against highly resistant mycobacteria, Gram-positive bacteria, some Gram-negative bacteria, and specific *Mycobacterium* species like *M. tuberculosis*, *M. leprae*, and *M. avium intracellulare*.
  • Inhibits DNA-dependent RNA polymerase.
  • Effective against highly resistant mycobacteria.
  • Interferes with HSP-90 molecular chaperone.
  • Enhances ubiquitination and protein degradation.
  • Inactivates bacterial RNA polymerase.
Catalog No. 50-003-84494 Supplier Medchemexpress LLC Supplier No. HY1702510MM/1ML
Rifabutin (Ansamycin) is a semisynthetic ansamycin antibiotic with potent antimycobacterial properties, primarily used as a bactericidal antibiotic to treat tuberculosis. It inhibits DNA-dependent RNA polymerase and is effective against highly resistant mycobacteria, Gram-positive bacteria, some Gram-negative bacteria, and specific *Mycobacterium* species like *M. tuberculosis*, *M. leprae*, and *M. avium intracellulare*.
  • Inhibits DNA-dependent RNA polymerase.
  • Effective against highly resistant mycobacteria.
  • Interferes with HSP-90 molecular chaperone.
  • Enhances ubiquitination and protein degradation.
  • Inactivates bacterial RNA polymerase.