Learn More
Medchemexpress LLC Rifabutin | 72559-06-9 | 99.2% | 847.00 | 25 MG
SDP
Catalog No. 5000385319
Encompass_Preferred
Rifabutin is a semisynthetic ansamycin antibiotic that exhibits potent antimycobacterial properties by inhibiting DNA-dependent RNA polymerase. It is primarily used as a bactericidal agent for treating tuberculosis and is effective against a range of resistant mycobacteria, including Mycobacterium tuberculosis, M. leprae, and M. avium intracellulare. It also acts as an antibiotic and antitumor agent, influencing HSP-90 molecular chaperone and inactivating bacterial RNA polymerase.
  • Semisynthetic ansamycin antibiotic
  • Inhibits DNA-dependent RNA polymerase
  • Bactericidal against resistant mycobacteria
  • Effective against Gram-positive and some Gram-negative bacteria
  • Treats tuberculosis and other mycobacterial infections
  • Acts as an antitumor agent
  • Modulates HSP-90 molecular chaperone
  • Inactivates bacterial RNA polymerase
Catalog No. 50-003-85319 Supplier Medchemexpress LLC Supplier No. 25MGHY17025
Rifabutin is a semisynthetic ansamycin antibiotic that exhibits potent antimycobacterial properties by inhibiting DNA-dependent RNA polymerase. It is primarily used as a bactericidal agent for treating tuberculosis and is effective against a range of resistant mycobacteria, including Mycobacterium tuberculosis, M. leprae, and M. avium intracellulare. It also acts as an antibiotic and antitumor agent, influencing HSP-90 molecular chaperone and inactivating bacterial RNA polymerase.
  • Semisynthetic ansamycin antibiotic
  • Inhibits DNA-dependent RNA polymerase
  • Bactericidal against resistant mycobacteria
  • Effective against Gram-positive and some Gram-negative bacteria
  • Treats tuberculosis and other mycobacterial infections
  • Acts as an antitumor agent
  • Modulates HSP-90 molecular chaperone
  • Inactivates bacterial RNA polymerase