Learn More
Medchemexpress LLC Rifabutin-d7 | 2747918-39-2 | 98.39% | 854.05 | 1 MG
SDP
Catalog No. 5000384666
Encompass_Preferred
Rifabutin-d7 is the deuterium labeled Rifabutin, a semisynthetic ansamycin antibiotic with potent antimycobacterial properties. It inhibits DNA-dependent RNA polymerase and is suitable for laboratory centrifugation and storage. The compound can be used for research purposes only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-84666 Supplier Medchemexpress LLC Supplier No. HY17025S1MG
Rifabutin-d7 is the deuterium labeled Rifabutin, a semisynthetic ansamycin antibiotic with potent antimycobacterial properties. It inhibits DNA-dependent RNA polymerase and is suitable for laboratory centrifugation and storage. The compound can be used for research purposes only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.