Learn More
Medchemexpress LLC Rifabutin (standard) | 72559-06-9 | 98.8% | 847.00 | 100 MG
SDP
Catalog No. 5000386068
Encompass_Preferred
Rifabutin (Standard) is the analytical standard of Rifabutin. This product is intended for research and analytical applications. Rifabutin (Ansamycin) is a semisynthetic ansamycin antibiotic with potent antimycobacterial properties, inhibiting DNA-dependent RNA polymerase. It is the grade of analytical standard, which is the reference standard supplied assay, commonly used in qualitative, quantitative, and methodological research experiments.
  • Inhibits DNA-dependent RNA polymerase
  • Suitable for HPLC, GC, and MS research experiments
  • Semisynthetic ansamycin antibiotic
  • Potent antimycobacterial properties
Catalog No. 50-003-86068 Supplier Medchemexpress LLC Supplier No. HY17025100MG
Rifabutin (Standard) is the analytical standard of Rifabutin. This product is intended for research and analytical applications. Rifabutin (Ansamycin) is a semisynthetic ansamycin antibiotic with potent antimycobacterial properties, inhibiting DNA-dependent RNA polymerase. It is the grade of analytical standard, which is the reference standard supplied assay, commonly used in qualitative, quantitative, and methodological research experiments.
  • Inhibits DNA-dependent RNA polymerase
  • Suitable for HPLC, GC, and MS research experiments
  • Semisynthetic ansamycin antibiotic
  • Potent antimycobacterial properties