Learn More
Medchemexpress LLC Rifabutin (Standard) | 72559-06-9 | 98.8% | 847.00 | 100 MG
SDP
Catalog No. 5000384295
Encompass_Preferred
Rifabutin (Standard) is the analytical standard of Rifabutin, intended for research and analytical applications. It is a semisynthetic ansamycin antibiotic with potent antimycobacterial properties, and it inhibits DNA-dependent RNA polymerase. The compound is of analytical standard grade, supplied as a reference standard assay, and is commonly used in qualitative, quantitative, and methodological research experiments.
  • Reference standard supplied for assay
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments in HPLC, GC, and MS
Catalog No. 50-003-84295 Supplier Medchemexpress LLC Supplier No. HY17025R100MG
Rifabutin (Standard) is the analytical standard of Rifabutin, intended for research and analytical applications. It is a semisynthetic ansamycin antibiotic with potent antimycobacterial properties, and it inhibits DNA-dependent RNA polymerase. The compound is of analytical standard grade, supplied as a reference standard assay, and is commonly used in qualitative, quantitative, and methodological research experiments.
  • Reference standard supplied for assay
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments in HPLC, GC, and MS