Learn More
Medchemexpress LLC Rifabutin (Standard) | 72559-06-9 | 98.8% | 847.00 | 50 MG
SDP
Catalog No. 5000385711
Encompass_Preferred
Rifabutin (Standard) is the analytical standard of Rifabutin, intended for research and analytical applications. It is a semisynthetic ansamycin antibiotic with potent antimycobacterial properties that inhibits DNA-dependent RNA polymerase.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.
  • Has potent antimycobacterial properties.
  • Inhibits DNA-dependent RNA polymerase.
Catalog No. 50-003-85711 Supplier Medchemexpress LLC Supplier No. HY17025R50MG
Rifabutin (Standard) is the analytical standard of Rifabutin, intended for research and analytical applications. It is a semisynthetic ansamycin antibiotic with potent antimycobacterial properties that inhibits DNA-dependent RNA polymerase.
  • Used in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.
  • Has potent antimycobacterial properties.
  • Inhibits DNA-dependent RNA polymerase.