Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Rifabutin (standard) | 72559-06-9 | MFCD00866816 | 98.8% | 847.00 g/mol | C46H62N4O11 | 10 MG
SDP

Catalog No. 5000295179
Encompass_Preferred

Rifabutin (Standard) is an analytical reference standard of the semisynthetic ansamycin antibiotic rifabutin. Supplied as a solid, it is intended for research and analytical applications, including HPLC, GC, and mass spectrometry, and is used in method development and quantitative assays.

  • High purity (98.8%).
  • Suitable for HPLC, GC, and mass spectrometry.
  • Solid formulation for analytical workflows.
  • Molecular formula C46H62N4O11; molecular weight 847.00 g/mol.
  • CAS number 72559-06-9.
  • Appearance brown to reddish brown.
  • Package size 10 MG.

Catalog No. 50-002-95179 Supplier Medchemexpress LLC Supplier No. HY17025R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rifabutin (Standard) is an analytical reference standard of the semisynthetic ansamycin antibiotic rifabutin. Supplied as a solid, it is intended for research and analytical applications, including HPLC, GC, and mass spectrometry, and is used in method development and quantitative assays.

  • High purity (98.8%).
  • Suitable for HPLC, GC, and mass spectrometry.
  • Solid formulation for analytical workflows.
  • Molecular formula C46H62N4O11; molecular weight 847.00 g/mol.
  • CAS number 72559-06-9.
  • Appearance brown to reddish brown.
  • Package size 10 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.