Learn More
Medchemexpress LLC Rifalazil 100mg | 129791-92-0 | 100MG
SDP
Catalog No. 5000243235
Encompass_Preferred
Rifalazil (KRM-1648 ABI-1648) a rifamycin derivative inhibits the bacterial DNA-dependent RNA polymerase and kills bacterial cells by blocking off the -subunit in RNA polymerase[1 Rifalazil (KRM-1648 ABI-1648) is an antibiotic exhibits high potency against mycobacteria gram-positive bacteria Helicobacter pylori C pneumoniae and C trachomatis with MIC values from 0 00025 to 0 0025 g/ml[3 Rifalazil (KRM-1648 ABI-1648) has the potential for the treatment of Chlamydia infection Clostridium difficile associated diarrhea (CDAD) and tuberculosis (TB)[2
Catalog No. 50-002-43235 Supplier Medchemexpress LLC Supplier No. HY105099100MG
Rifalazil (KRM-1648 ABI-1648) a rifamycin derivative inhibits the bacterial DNA-dependent RNA polymerase and kills bacterial cells by blocking off the -subunit in RNA polymerase[1 Rifalazil (KRM-1648 ABI-1648) is an antibiotic exhibits high potency against mycobacteria gram-positive bacteria Helicobacter pylori C pneumoniae and C trachomatis with MIC values from 0 00025 to 0 0025 g/ml[3 Rifalazil (KRM-1648 ABI-1648) has the potential for the treatment of Chlamydia infection Clostridium difficile associated diarrhea (CDAD) and tuberculosis (TB)[2