Learn More
Medchemexpress LLC Rifalazil | 129791-92-0 | 99.6% | 941.07 | C51H64N4O13 | 50 MG
SDP
Catalog No. 5000244365
Encompass_Preferred
Rifalazil is a rifamycin-class antibiotic research compound that inhibits bacterial DNA-dependent RNA polymerase by binding the β-subunit. It demonstrates potent activity against a range of pathogens and is provided for research use only.
  • Inhibits bacterial RNA polymerase by targeting the β-subunit.
  • Exhibits high potency against mycobacteria and gram-positive bacteria.
  • Active against Helicobacter pylori, Chlamydia pneumoniae, and Chlamydia trachomatis.
  • High purity (~99.6%) suitable for research applications.
  • Available as a 50 MG solid package.
  • Stable when stored sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Intended for research use only.
Catalog No. 50-002-44365 Supplier Medchemexpress LLC Supplier No. HY10509950MG
Rifalazil is a rifamycin-class antibiotic research compound that inhibits bacterial DNA-dependent RNA polymerase by binding the β-subunit. It demonstrates potent activity against a range of pathogens and is provided for research use only.
  • Inhibits bacterial RNA polymerase by targeting the β-subunit.
  • Exhibits high potency against mycobacteria and gram-positive bacteria.
  • Active against Helicobacter pylori, Chlamydia pneumoniae, and Chlamydia trachomatis.
  • High purity (~99.6%) suitable for research applications.
  • Available as a 50 MG solid package.
  • Stable when stored sealed at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Intended for research use only.