Learn More
Enzo Life Sciences Rifampicin (100mg). CAS: 13292-46-1
SDP
Supplier:  Enzo Life Sciences ALX380071M100
Encompass_Preferred
A selective inhibitor of bacterial DNA-dependent RNA polymerase (RNAP). Has no effect on mammalian RNA polymerase. The RNAP-rifampicin complex locks the promoter in an abortive initiation reaction making short oligoribonucleotides which diffuse out of the active site. Inhibits α-synuclein fibrillation and disaggregates fibrils. Potent inducer of hepatic cytochrome P450 enzymes. Alternative name: Rifampin, 3-(4-Methylpiperazinyl-iminomethyl)rifamycin SV. Purity: ≥97% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or methanol (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-8945