Learn More
Medchemexpress LLC Rifampicin | 13292-46-1 | 98.2% | 1 ML
SDP
Catalog No. 5000405986
Encompass_Preferred
Rifampicin is a potent, broad-spectrum antibiotic effective against bacterial pathogens, and it also exhibits anti-influenza virus and anti-orthopoxvirus activities. It is intended for research use only.
  • Potent, broad-spectrum antibiotic
  • Effective against bacterial pathogens
  • Exhibits anti-influenza virus activity
  • Exhibits anti-orthopoxvirus activity
  • Available as a 10 mM * 1 mL solution in DMSO
  • Ready for reconstitution
Catalog No. 50-004-05986 Supplier Medchemexpress LLC Supplier No. HYB027210MM/1ML
Rifampicin is a potent, broad-spectrum antibiotic effective against bacterial pathogens, and it also exhibits anti-influenza virus and anti-orthopoxvirus activities. It is intended for research use only.
  • Potent, broad-spectrum antibiotic
  • Effective against bacterial pathogens
  • Exhibits anti-influenza virus activity
  • Exhibits anti-orthopoxvirus activity
  • Available as a 10 mM * 1 mL solution in DMSO
  • Ready for reconstitution