Learn More
Medchemexpress LLC HY-B0272 1g Medchemexpress, Rifampicin CAS:13292-46-1 Purity:>98%
SDP
Catalog No. 502029396
Encompass_Preferred
Medchemexpress, HY-B0272 1g Rifampicin CAS:13292-46-1 Rifampicin is a potent and broad spectrum antibiotic against bacterial pathogens. Rifampicin has anti-influenza virus activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9396 Supplier Medchemexpress LLC Supplier No. HYB02721G
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-B0272 1g Rifampicin CAS:13292-46-1 Rifampicin is a potent and broad spectrum antibiotic against bacterial pathogens. Rifampicin has anti-influenza virus activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.