Learn More
Cayman Chemical RIfampIcIn-d3 500ug
SDP
Catalog No. 502939427
Encompass_Preferred
An internal standard for the quantification of rifampicin by GC- or LC-MS
Catalog No. 50-293-9427 Supplier Cayman Chemical Supplier No. 27848500 UG
An internal standard for the quantification of rifampicin by GC- or LC-MS