Learn More
Medchemexpress LLC Rifampicin-d4 | 2747918-60-9 | 99.7% | 1 MG
SDP
Catalog No. 5000408105
Encompass_Preferred
Rifampicin-d4 is a deuterium-labeled form of Rifampicin, a potent, broad-spectrum antibiotic with activity against bacterial pathogens and influenza viruses. It is intended for research use only.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporation of deuterium allows for quantitation during drug development.
  • Deuteration can influence pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-08105 Supplier Medchemexpress LLC Supplier No. HYB0272S21MG
Rifampicin-d4 is a deuterium-labeled form of Rifampicin, a potent, broad-spectrum antibiotic with activity against bacterial pathogens and influenza viruses. It is intended for research use only.
  • Can be used as a tracer.
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporation of deuterium allows for quantitation during drug development.
  • Deuteration can influence pharmacokinetic and metabolic profiles of drugs.