Learn More
Crescent Chemical Co Inc RIFAMPICIN RESEARCH GRADE
SDP
Catalog No. 50248106
Encompass_Preferred
Rifampicin research grade Semisynthetic derivative of rifamycin SV which is produced from certain strains of Amycolatopsis mediterranei. Belongs to the group of ansamycin antibiotics. Inhibitor of DNA-dependent RNA-polymerase. Specifically inhibits the initi
Catalog No. 50-248-106 Supplier Crescent Chemical Co Inc Supplier No. SE34514.01
Rifampicin research grade Semisynthetic derivative of rifamycin SV which is produced from certain strains of Amycolatopsis mediterranei. Belongs to the group of ansamycin antibiotics. Inhibitor of DNA-dependent RNA-polymerase. Specifically inhibits the initi