Learn More
Medchemexpress LLC Rifampicin (Standard) | 13292-46-1 | 95.5% | 100 MG
SDP
Catalog No. 5000408713
Encompass_Preferred
Rifampicin (Standard) is the analytical standard of Rifampicin, intended for research and analytical applications. It is a potent, broad-spectrum antibiotic effective against bacterial pathogens. Rifampicin also exhibits anti-influenza virus and anti-orthopoxvirus activities.
  • Analytical standard grade for assay reference
  • Used in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS
  • Potent and broad-spectrum antibiotic against bacterial pathogens
  • Exhibits anti-influenza virus activities
  • Shows anti-orthopoxvirus activity
Catalog No. 50-004-08713 Supplier Medchemexpress LLC Supplier No. HYB0272R100MG
Rifampicin (Standard) is the analytical standard of Rifampicin, intended for research and analytical applications. It is a potent, broad-spectrum antibiotic effective against bacterial pathogens. Rifampicin also exhibits anti-influenza virus and anti-orthopoxvirus activities.
  • Analytical standard grade for assay reference
  • Used in qualitative, quantitative, and methodological research experiments involving HPLC, GC, and MS
  • Potent and broad-spectrum antibiotic against bacterial pathogens
  • Exhibits anti-influenza virus activities
  • Shows anti-orthopoxvirus activity