Learn More
Medchemexpress LLC Rifampicin (Standard) | 13292-46-1 | 95.5% | 25 MG
SDP
Catalog No. 5000450629
Encompass_Preferred
Rifampicin (Standard) is a potent and broad-spectrum antibiotic effective against bacterial pathogens. It also exhibits anti-influenza virus and anti-orthopoxvirus activities. This analytical standard is intended for research and analytical applications.
  • Used as a reference standard for assays
  • Employed in qualitative, quantitative, and methodological research experiments
  • Effective against bacterial pathogens
  • Exhibits anti-influenza virus activity
  • Shows anti-orthopoxvirus activity
Catalog No. 50-004-50629 Supplier Medchemexpress LLC Supplier No. 25MGHYB0272R
Rifampicin (Standard) is a potent and broad-spectrum antibiotic effective against bacterial pathogens. It also exhibits anti-influenza virus and anti-orthopoxvirus activities. This analytical standard is intended for research and analytical applications.
  • Used as a reference standard for assays
  • Employed in qualitative, quantitative, and methodological research experiments
  • Effective against bacterial pathogens
  • Exhibits anti-influenza virus activity
  • Shows anti-orthopoxvirus activity