Learn More
Medchemexpress LLC Rifampicin (Standard) | 13292-46-1 | 95.5% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0272R50MG
Encompass_Preferred
Rifampicin (Standard) is the analytical standard of Rifampicin, intended for research and analytical applications. It is a potent and broad-spectrum antibiotic effective against bacterial pathogens, and also exhibits anti-influenza virus and anti-orthopoxvirus activities.
  • Analytical standard for Rifampicin
  • Potent and broad-spectrum antibiotic
  • Exhibits anti-influenza virus activities
  • Shows anti-orthopoxvirus activity
  • Reference standard supplied assay
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
Catalog No. 50-004-06024