Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rifampicin 98.0+%, TCI America™
SDP

Catalog No. R007925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R007925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R007925G Supplier TCI America Supplier No. R007925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An ansamycin antibiotic reagent for research use, with a purity of >98.0%.

Specifications

Type Rifampicin
Physical Form Crystalline Powder
CAS 13292-46-1
Melting Point 188°C
Color Orange
Quantity 25 g
Molecular Formula C43H58N4O12
MDL Number MFCD00151389
Synonym Rifampin, Rifamycin AMP
InChI Key FZYOVNIOYYPUPY-HRJPTAQKSA-N
SMILES CC1C=CC=C(C(=O)NC2=C(C3=C(C(=C4C(=C3C(=O)C2=CNN5CCN(CC5)C)C(=O)C(O4)(OC=CC(C(C(C(C(C(C1O)C)O)C)OC(=O)C)C)OC)C)C)O)O)C
Molecular Weight (g/mol) 822.953
PubChem CID 131839595
Formula Weight 822.95
Percent Purity ≥98.0% (T)
Grade Not Available
Chemical Name or Material Rifampicin
Show More Show Less
EINECSNumber (8)-0463
RTECSNumber VJ7000000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.