Learn More
Apexbio Technology LLC Rifampin(Synonyms: Rifampicin, Rifaldazine, Rifadin, Rimactane, Rifoldin, Rofact, Rifamycin AMP, Riforal), 10mM (in 1mL DMSO), CAS: 13292-46-1.
SDP
Catalog No. 5000060572
Encompass_Preferred
Rifampin (CAS 13292-46-1) is an antibiotic belonging to the rifamycin family characterized by bactericidal activity via selective inhibition of bacterial DNA-dependent RNA polymerase This inhibition disrupts transcriptional initiation by preventing RNA synthesis leading to impaired bacterial protein biosynthesis and subsequent cell death In biomedical research rifampin serves as an essential tool for investigations into bacterial resistance mechanisms transcriptional regulation and synthetic biology studies that rely on specific transcription inhibition
Catalog No. 50-000-60572 Supplier Apexbio Technology LLC Supplier No. B20215.1
Rifampin (CAS 13292-46-1) is an antibiotic belonging to the rifamycin family characterized by bactericidal activity via selective inhibition of bacterial DNA-dependent RNA polymerase This inhibition disrupts transcriptional initiation by preventing RNA synthesis leading to impaired bacterial protein biosynthesis and subsequent cell death In biomedical research rifampin serves as an essential tool for investigations into bacterial resistance mechanisms transcriptional regulation and synthetic biology studies that rely on specific transcription inhibition