Learn More
Apexbio Technology LLC Rifampin(Synonyms: Rifampicin, Rifaldazine, Rifadin, Rimactane, Rifoldin, Rofact, Rifamycin AMP, Riforal), 5g, CAS: 13292-46-1.
SDP
Catalog No. 5000060550
Encompass_Preferred
Rifampin (CAS 13292-46-1) is an antibiotic belonging to the rifamycin family characterized by bactericidal activity via selective inhibition of bacterial DNA-dependent RNA polymerase This inhibition disrupts transcriptional initiation by preventing RNA synthesis leading to impaired bacterial protein biosynthesis and subsequent cell death In biomedical research rifampin serves as an essential tool for investigations into bacterial resistance mechanisms transcriptional regulation and synthetic biology studies that rely on specific transcription inhibition
Catalog No. 50-000-60550 Supplier Apexbio Technology LLC Supplier No. B20215000
Rifampin (CAS 13292-46-1) is an antibiotic belonging to the rifamycin family characterized by bactericidal activity via selective inhibition of bacterial DNA-dependent RNA polymerase This inhibition disrupts transcriptional initiation by preventing RNA synthesis leading to impaired bacterial protein biosynthesis and subsequent cell death In biomedical research rifampin serves as an essential tool for investigations into bacterial resistance mechanisms transcriptional regulation and synthetic biology studies that rely on specific transcription inhibition